Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP89A2
- Entrez Gene ID
-
842798
- Gene Name
- cytochrome P450, family 89, subfamily A, polypeptide 2
- Gene Aliases
-
''cytochrome P450, AT1G64900, CYP89, F13O11.20, F13O11_20, cytochrome P450, family 89, polypeptide 2, polypeptide 2'', subfamily A
- Chromosome Location
-
Chr.1: 24113109 - 24114984 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105164.3 |
NP_176670.1 |
1 - 1 |
1754 |
119 |
GenBank mRNA |
BT002531.1 |
- |
1 - 1 |
1647 |
119 |
View Full Product Details