Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRAMP5
Entrez Gene ID
827613
Gene Name
NRAMP metal ion transporter family protein
Gene Aliases
AT4G18790, ATNRAMP5, F28A21.200, F28A21_200
Chromosome Location
Chr.4: 10317416 - 10319736 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117995.2 NP_193614.1 1 - 2 653 138
GenBank mRNA AJ292076.1 - 1 - 2 577 138

View Full Product Details