Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HAC2
- Entrez Gene ID
-
843042
- Gene Name
- histone acetyltransferase of the CBP family 2
- Gene Aliases
-
ARABIDOPSIS THALIANA P300/CBP ACETYLTRANSFERASE-RELATED PROTEIN 1, AT1G67220, ATHPCAT1, F1N21.4, HAC02, histone acetyltransferase of the CBP family 2
- Chromosome Location
-
Chr.1: 25145587 - 25150558 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105391.4 |
NP_564891.4 |
4 - 5 |
2787 |
80 |
View Full Product Details