Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLH3
Entrez Gene ID
829704
Gene Name
MUTL protein homolog 3
Gene Aliases
AT4G35520, ATMLH3, F8D20.30, F8D20_30, MUTL protein homolog 3
Chromosome Location
Chr.4: 16865304 - 16871527 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342354.1 NP_001320143.1 14 - 15 2633 65
NM_001342355.1 NP_001328252.1 15 - 16 2892 65
NM_001342356.1 NP_001328253.1 14 - 15 2811 65
NM_001342357.1 NP_001328256.1 14 - 15 2633 65
NM_001342358.1 NP_001328251.1 14 - 15 2597 65
NM_001342359.1 NP_001328254.1 14 - 15 2597 65
NM_001342360.1 NP_001328255.1 6 - 7 1971 65
NM_119717.7 NP_195277.6 6 - 7 2033 65

View Full Product Details