Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAPRT1
Entrez Gene ID
829848
Gene Name
nicotinate phosphoribosyltransferase 1
Gene Aliases
AP22.31, AP22_31, AT4G36940, nicotinate phosphoribosyltransferase 1
Chromosome Location
Chr.4: 17416855 - 17420252 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119858.6 NP_195412.3 10 - 11 1231 85
GenBank mRNA AY093004.1 - 10 - 11 1199 85

View Full Product Details