Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEA
- Entrez Gene ID
-
839422
- Gene Name
- SET domain-containing protein
- Gene Aliases
-
AT1G02580, EMB173, EMBRYO DEFECTIVE 173, FERTILIZATION INDEPENDENT SEED 1, FIS1, MEDEA, SDG5, SET DOMAIN-CONTAINING PROTEIN 5, T14P4.11, T14P4_11
- Chromosome Location
-
Chr.1: 544733 - 549202 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100139.4 |
NP_563658.1 |
14 - 15 |
1746 |
156 |
GenBank mRNA |
AF060485.1 |
- |
14 - 15 |
1696 |
156 |
View Full Product Details