Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOE2
Entrez Gene ID
836134
Gene Name
target of early activation tagged (EAT) 2
Gene Aliases
AT5G60120, MGO3.10, MGO3_10, target of early activation tagged (EAT) 2
Chromosome Location
Chr.5: 24207689 - 24211743 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203647.1 NP_001190576.1 3 - 4 1055 99
NM_001345390.1 NP_001331668.1 3 - 4 1055 99
NM_001345391.1 NP_001331666.1 3 - 4 1055 99
NM_001345392.1 NP_001331665.1 3 - 4 1055 99
NM_001345393.1 NP_001331667.1 3 - 4 1055 99
NM_125405.4 NP_200820.3 3 - 4 1055 99
GenBank mRNA BT001916.1 - 3 - 4 591 99

View Full Product Details