Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYS1
Entrez Gene ID
831087
Gene Name
cystatin-1
Gene Aliases
AT5G12140, ATCYS1, MXC9.10, MXC9_10, cystatin-1
Chromosome Location
Chr.5: 3922591 - 3924058 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121252.3 NP_196775.1 1 - 2 242 69
GenBank mRNA AY088332.1 - 1 - 2 207 69

View Full Product Details