Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATXR2
Entrez Gene ID
821736
Gene Name
histone-lysine N-methyltransferase ATXR2
Gene Aliases
AT3G21820, SDG36, SET DOMAIN PROTEIN 36, histone-lysine N-methyltransferase ATXR2
Chromosome Location
Chr.3: 7688434 - 7691567 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113077.5 NP_188819.2 13 - 14 1424 81
GenBank mRNA AK221711.1 - 13 - 14 1361 81
BT020260.1 - 13 - 14 1308 81
BT021112.1 - 13 - 14 1301 81

View Full Product Details