Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl21
Entrez Gene ID
822126
Gene Name
serine carboxypeptidase-like 21
Gene Aliases
AT3G25420, serine carboxypeptidase-like 21
Chromosome Location
Chr.3: 9217832 - 9220836 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338744.1 NP_001319641.1 8 - 9 1048 60

View Full Product Details