Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRO4
Entrez Gene ID
823926
Gene Name
similar to RCD one 4
Gene Aliases
AT3G47720, T23J7_50, similar to RCD one 4
Chromosome Location
Chr.3: 17592558 - 17594132 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114640.4 NP_190356.2 2 - 3 607 135

View Full Product Details