Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA8
Entrez Gene ID
823933
Gene Name
ABC2 homolog 7
Gene Aliases
ABC2 homolog 7, AT3G47790, ATATH7, ATH7, ATP-binding cassette A8
Chromosome Location
Chr.3: 17629530 - 17633942 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339337.1 NP_001327142.1 2 - 3 592 213
NM_001339338.1 NP_001327141.1 3 - 4 501 122
NM_114647.4 NP_190363.3 3 - 4 501 122
GenBank mRNA AY099665.1 - 3 - 4 500 122

View Full Product Details