Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPRO2
Entrez Gene ID
818588
Gene Name
ortholog of sugar beet HS1 PRO-1 2
Gene Aliases
ARABIDOPSIS ORTHOLOG OF SUGAR BEET HS1 PRO-1 2, AT2G40000, ATHSPRO2, T28M21.16, T28M21_16, ortholog of sugar beet HS1 PRO-1 2
Chromosome Location
Chr.2: 16700490 - 16702336 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129558.3 NP_181529.1 1 - 1 1570 87
GenBank mRNA AY080778.1 - 1 - 1 1260 87
AY117156.1 - 1 - 1 1190 87
DQ132635.1 - 1 - 1 1259 87

View Full Product Details