Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BNIP2
Entrez Gene ID
516077
Gene Name
BCL2 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 50557184 - 50578063 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040534.2 NP_001035624.2 4 - 5 545 116
GenBank mRNA BT021775.1 - 4 - 5 492 116
EE236937.1 - 4 - 5 546 116

View Full Product Details