Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FXYD6
Entrez Gene ID
511064
Gene Name
FXYD domain containing ion transport regulator 6
Gene Aliases
-
Chromosome Location
Chr.15: 28497530 - 28534963 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034399.1 NP_001029571.1 4 - 5 214 77
GenBank mRNA BC104508.1 - 4 - 5 214 77

View Full Product Details