Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTCAP3
Entrez Gene ID
508550
Gene Name
keratinocyte associated protein 3
Gene Aliases
-
Chromosome Location
Chr.11: 72232725 - 72234438 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035062.2 NP_001030234.1 3 - 4 304 86
GenBank mRNA BC103079.1 - 3 - 4 305 86

View Full Product Details