Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
506760
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
-
Chromosome Location
Chr.25: 7685029 - 7698191 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045951.1 NP_001039416.1 3 - 4 328 69
GenBank mRNA BC111637.1 - 3 - 4 328 69

View Full Product Details