Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISOC2
Entrez Gene ID
516212
Gene Name
isochorismatase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 62053772 - 62060244 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038123.2 NP_001033212.1 1 - 2 68 138
GenBank mRNA BC109878.1 - 1 - 2 69 138

View Full Product Details