Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP2
Entrez Gene ID
532432
Gene Name
receptor accessory protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 49807954 - 49814912 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046325.2 NP_001039790.1 3 - 4 282 80
GenBank mRNA BC112790.1 - 3 - 4 283 80

View Full Product Details