Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NONO
Entrez Gene ID
615175
Gene Name
non-POU domain containing octamer binding
Gene Aliases
-
Chromosome Location
Chr.X: 79368090 - 79381393 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046554.1 NP_001040019.1 6 - 7 993 78
GenBank mRNA BC105532.1 - 6 - 7 993 78

View Full Product Details