Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBGT1
Entrez Gene ID
618517
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.11: 103136834 - 103147285 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076509.2 NP_001069977.1 4 - 5 360 101
GenBank mRNA BC118408.1 - 4 - 5 361 101

View Full Product Details