Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP3A5
Entrez Gene ID
282214
Gene Name
cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4
Gene Aliases
CYP3A28, CYP3A4
Chromosome Location
Chr.25: 36530333 - 36568936 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174531.3 NP_776956.2 12 - 13 1496 73
GenBank mRNA BC114003.1 - 12 - 13 1497 73
Y10214.1 - - 1415 73

View Full Product Details