Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TMEM65
Entrez Gene ID
614243
Gene Name
transmembrane protein 65
Gene Aliases
-
Chromosome Location
Chr.14: 15674944 - 15722010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076318.1 NP_001069786.1 1 - 2 659 105
GenBank mRNA BC120160.1 - 1 - 2 659 105

View Full Product Details