Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPANK1
Entrez Gene ID
617553
Gene Name
G-patch domain and ankyrin repeats 1
Gene Aliases
BAT4
Chromosome Location
Chr.23: 27634113 - 27637417 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076468.1 NP_001069936.1 1 - 2 214 105
GenBank mRNA BC120141.1 - 1 - 2 214 105

View Full Product Details