Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEANC
Entrez Gene ID
504389
Gene Name
transcription elongation factor A N-terminal and central domain containing
Gene Aliases
-
Chromosome Location
Chr.X: 129855107 - 129866216 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024472.2 NP_001019643.2 2 - 3 139 69
GenBank mRNA BC126748.1 - 2 - 3 139 69
BT021661.1 - 2 - 3 138 69

View Full Product Details