Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC20
Entrez Gene ID
521721
Gene Name
leucine rich repeat containing 20
Gene Aliases
-
Chromosome Location
Chr.28: 26507230 - 26577673 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100387.1 NP_001093857.1 1 - 2 68 56
GenBank mRNA BC147923.1 - 1 - 2 68 56

View Full Product Details