Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBY2
Entrez Gene ID
616053
Gene Name
chibby family member 2
Gene Aliases
SPERT
Chromosome Location
Chr.12: 15768622 - 15782386 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102297.2 NP_001095767.1 1 - 2 80 72
GenBank mRNA BC149972.1 - 1 - 2 81 72

View Full Product Details