Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PADI2
Entrez Gene ID
617851
Gene Name
peptidyl arginine deiminase 2
Gene Aliases
-
Chromosome Location
Chr.2: 135473392 - 135529995 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105452.2 NP_001098922.1 11 - 12 1392 75
GenBank mRNA BC149017.1 - 11 - 12 1393 75

View Full Product Details