Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT5B
Entrez Gene ID
508008
Gene Name
Wnt family member 5B
Gene Aliases
-
Chromosome Location
Chr.5: 108071224 - 108151542 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205628.1 NP_001192557.1 3 - 4 614 62

View Full Product Details