Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAT1
Entrez Gene ID
614942
Gene Name
T cell receptor associated transmembrane adaptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 53562091 - 53626467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163943.1 NP_001157415.1 2 - 3 257 93
GenBank mRNA BC103345.1 - 2 - 3 257 93

View Full Product Details