Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT15
Entrez Gene ID
618078
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNTL2
Chromosome Location
Chr.1: 153179306 - 153226107 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193182.1 NP_001180111.1 8 - 9 1622 72

View Full Product Details