Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TASOR
Entrez Gene ID
510095
Gene Name
transcription activation suppressor
Gene Aliases
C22H3orf63, FAM208A
Chromosome Location
Chr.22: 44352988 - 44404518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205393.2 NP_001192322.1 21 - 22 4023 99
GenBank mRNA BC151347.1 - 13 - 14 2901 99

View Full Product Details