Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TASOR
Entrez Gene ID
510095
Gene Name
transcription activation suppressor
Gene Aliases
C22H3orf63, FAM208A
Chromosome Location
Chr.22: 44352988 - 44404518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205393.2 NP_001192322.1 9 - 10 1157 98

View Full Product Details