Product Details

SNP ID
rs79827771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104605009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATTAATAACAAAGGTGTCAATA[T/G]GAACAATGTTGTGGCCACAAGCATG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
OR13C2 PubMed Links

Gene Details

Gene
OR13C2
Gene Name
olfactory receptor family 13 subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004481.1 619 Missense Mutation GTA,TTA V207L NP_001004481.1
Gene
OR13C5
Gene Name
olfactory receptor family 13 subfamily C member 5
There are no transcripts associated with this gene.

View Full Product Details