Product Details

SNP ID
rs79449664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:842425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGCGCCTCACCCCTCGGTGTA[C/T]TCGGCCTGCAGCAGCCCCAGGTAAG
Phenotype
MIM: 602243 MIM: 601189 MIM: 602644
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD151 PubMed Links

Gene Details

Gene
CD151
Gene Name
CD151 molecule (Raph blood group)
There are no transcripts associated with this gene.

Gene
POLR2L
Gene Name
RNA polymerase II subunit L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021128.4 105 Silent Mutation GAA,GAG E28E NP_066951.1
Gene
TSPAN4
Gene Name
tetraspanin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025234.1 105 Intron NP_001020405.1
NM_001025235.1 105 Intron NP_001020406.1
NM_001025236.1 105 Intron NP_001020407.1
NM_001025237.1 105 Intron NP_001020408.1
NM_001025238.1 105 Intron NP_001020409.1
NM_001025239.1 105 Intron NP_001020410.1
NM_003271.4 105 Intron NP_003262.1
XM_005253102.1 105 Intron XP_005253159.1
XM_005253104.1 105 Intron XP_005253161.1
XM_011520336.1 105 Intron XP_011518638.1
XM_011520337.2 105 Intron XP_011518639.1
XM_011520338.1 105 Intron XP_011518640.1
XM_011520339.2 105 Intron XP_011518641.1
XM_011520340.2 105 Intron XP_011518642.1
XM_011520341.2 105 Intron XP_011518643.1

View Full Product Details