Product Details

SNP ID
rs74345847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41216448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGCTTTACTGTCTGTACCCAG[A/C]TGCTCATTGTTATTGAATGACTCAT
Phenotype
MIM: 604601
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929140 PubMed Links

Gene Details

Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

Gene
MTRF1
Gene Name
mitochondrial translational release factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004294.2 2416 UTR 3 NP_004285.2
XM_005266599.2 2416 Intron XP_005266656.1
XM_006719897.2 2416 Intron XP_006719960.1
XM_006719898.2 2416 Intron XP_006719961.1
XM_006719900.2 2416 Intron XP_006719963.1
XM_011535315.2 2416 Intron XP_011533617.1
XM_011535317.2 2416 Intron XP_011533619.1
XM_011535318.2 2416 Intron XP_011533620.1
XM_011535320.2 2416 Intron XP_011533622.1
XM_011535321.2 2416 Intron XP_011533623.1
XM_011535323.2 2416 Intron XP_011533625.1
XM_011535325.2 2416 Intron XP_011533627.1
XM_011535326.2 2416 Intron XP_011533628.1
XM_011535327.2 2416 Intron XP_011533629.1
XM_017020856.1 2416 UTR 3 XP_016876345.1
XM_017020857.1 2416 Intron XP_016876346.1
XM_017020858.1 2416 Intron XP_016876347.1
XM_017020859.1 2416 Intron XP_016876348.1
XM_017020860.1 2416 Intron XP_016876349.1
XM_017020861.1 2416 Intron XP_016876350.1
XM_017020862.1 2416 Intron XP_016876351.1
XM_017020863.1 2416 Intron XP_016876352.1
XM_017020864.1 2416 Intron XP_016876353.1
XM_017020865.1 2416 Intron XP_016876354.1
XM_017020866.1 2416 Intron XP_016876355.1
XM_017020867.1 2416 Intron XP_016876356.1
XM_017020868.1 2416 Intron XP_016876357.1
XM_017020869.1 2416 Intron XP_016876358.1
XM_017020870.1 2416 Intron XP_016876359.1
XM_017020871.1 2416 Intron XP_016876360.1
XM_017020872.1 2416 Intron XP_016876361.1
XM_017020873.1 2416 Intron XP_016876362.1
XM_017020874.1 2416 Intron XP_016876363.1
XM_017020875.1 2416 Intron XP_016876364.1
XM_017020876.1 2416 Intron XP_016876365.1
XM_017020877.1 2416 Intron XP_016876366.1
XM_017020878.1 2416 Intron XP_016876367.1
XM_017020879.1 2416 Intron XP_016876368.1

View Full Product Details