Product Details

SNP ID
rs74337849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79682538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACACAATTTACAGCTAACCCTC[A/G]AACAACTTGCTTTGGTCTGCGAGGG
Phenotype
MIM: 604927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDP1 PubMed Links

Gene Details

Gene
CTDP1
Gene Name
CTD phosphatase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202504.1 303 Intron NP_001189433.1
NM_001318511.1 303 Intron NP_001305440.1
NM_004715.4 303 Intron NP_004706.3
NM_048368.3 303 Intron NP_430255.2
XM_011526261.1 303 Intron XP_011524563.1
XM_017026078.1 303 Intron XP_016881567.1
Gene
LOC105376875
Gene Name
uncharacterized LOC105376875
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011526290.2 303 Intron XP_011524592.1
XM_011526292.2 303 UTR 5 XP_011524594.1
XM_017026112.1 303 Intron XP_016881601.1
Gene
LOC284241
Gene Name
uncharacterized LOC284241
There are no transcripts associated with this gene.

View Full Product Details