Product Details

SNP ID
rs76271227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85804953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGAGTTCATATTCTTTTGCTT[T/G]CATCTGAAGCAATGAGTCACTGTAT
Phenotype
MIM: 610383
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
RASSF9 PubMed Links

Gene Details

Gene
RASSF9
Gene Name
Ras association domain family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005447.3 1396 Missense Mutation AAA,CAA K353Q NP_005438.2
XM_011538988.2 1396 Missense Mutation AAA,CAA K332Q XP_011537290.1

View Full Product Details