Product Details

SNP ID
rs78679797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118054029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTAGGATATCACTGAACTCTT[C/T]TGCCTTCATAACCAAATGTCAAAAT
Phenotype
MIM: 608598 MIM: 608599
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASC2 PubMed Links

Gene Details

Gene
CASC2
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RAB11FIP2
Gene Name
RAB11 family interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details