Product Details

SNP ID
rs76814823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118060302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGAGTAACTGGATGAGCTTGGG[A/C]AGTGTGACATAATAGCTTTGATCAG
Phenotype
MIM: 608598
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASC2 PubMed Links

Gene Details

Gene
CASC2
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details