Product Details

SNP ID
rs75699479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59833751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCATCTTTTCTTTGAAAAAGA[A/G]AAGATGCTTTTTTCCTTTCCATGAC
Phenotype
MIM: 609342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 Intron NP_005133.2
XM_011544939.2 Intron XP_011543241.1

View Full Product Details