Product Details

SNP ID
rs116164766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:88195980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCTCATACCAGTAAATACAAA[C/G]AGTTAAAATTCCAATGCCACAGTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM161B PubMed Links

Gene Details

Gene
TMEM161B
Gene Name
transmembrane protein 161B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289007.1 1638 Intron NP_001275936.1
NM_001289008.1 1638 UTR 3 NP_001275937.1
NM_153354.4 1638 Intron NP_699185.1
XM_006714553.1 1638 Intron XP_006714616.1
XM_006714554.3 1638 Intron XP_006714617.1
XM_006714555.3 1638 Intron XP_006714618.1
XM_006714556.3 1638 Intron XP_006714619.2
XM_011543201.2 1638 Intron XP_011541503.1
XM_011543202.2 1638 Intron XP_011541504.2
XM_011543203.2 1638 Intron XP_011541505.2
XM_011543204.2 1638 Intron XP_011541506.2
XM_017009093.1 1638 Intron XP_016864582.1
XM_017009094.1 1638 Intron XP_016864583.1
XM_017009095.1 1638 Intron XP_016864584.1
XM_017009096.1 1638 Intron XP_016864585.1
XM_017009097.1 1638 Intron XP_016864586.1
XM_017009098.1 1638 Intron XP_016864587.1
XM_017009099.1 1638 Intron XP_016864588.1
XM_017009100.1 1638 Intron XP_016864589.1
XM_017009101.1 1638 Intron XP_016864590.1
XM_017009102.1 1638 Intron XP_016864591.1
XM_017009103.1 1638 Intron XP_016864592.1
XM_017009104.1 1638 Intron XP_016864593.1
XM_017009105.1 1638 Intron XP_016864594.1
XM_017009106.1 1638 Intron XP_016864595.1
XM_017009107.1 1638 Intron XP_016864596.1
XM_017009108.1 1638 Intron XP_016864597.1
XM_017009109.1 1638 UTR 3 XP_016864598.1
XM_017009110.1 1638 UTR 3 XP_016864599.1

View Full Product Details