Product Details

SNP ID
rs116781812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30095764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTTTCGGGGGAGCATGGGCGG[A/G]TAGGCACTGGTTGGTCCTTGCGACC
Phenotype
MIM: 616318 MIM: 602427 MIM: 609724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD3 PubMed Links

Gene Details

Gene
GDPD3
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
There are no transcripts associated with this gene.

Gene
LOC101928595
Gene Name
uncharacterized LOC101928595
There are no transcripts associated with this gene.

Gene
TBX6
Gene Name
T-box 6
There are no transcripts associated with this gene.

Gene
YPEL3
Gene Name
yippee like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145524.1 437 Intron NP_001138996.1
NM_031477.4 437 UTR 5 NP_113665.3

View Full Product Details