Product Details

SNP ID
rs117438485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2768098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCTACTGAAGGGTGCATGGGT[C/T]GCCCTCAGTCTTTTGCTATTATAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAP1GAP2 PubMed Links

Gene Details

Gene
RAP1GAP2
Gene Name
RAP1 GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100398.1 Intron NP_001093868.1
NM_015085.4 Intron NP_055900.4
XM_005256542.2 Intron XP_005256599.1
XM_006721477.3 Intron XP_006721540.1
XM_011523738.1 Intron XP_011522040.1
XM_011523739.1 Intron XP_011522041.1
XM_011523740.2 Intron XP_011522042.1
XM_011523741.2 Intron XP_011522043.1
XM_011523742.2 Intron XP_011522044.1
XM_011523743.2 Intron XP_011522045.1
XM_011523744.2 Intron XP_011522046.1
XM_011523745.2 Intron XP_011522047.1
XM_017024370.1 Intron XP_016879859.1
XM_017024371.1 Intron XP_016879860.1

View Full Product Details