Product Details

SNP ID
rs115391024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35291412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAAATCTGGCGATCAAATTCT[C/T]ATGCAGCTCTCTCCACTTTTTTAAG
Phenotype
MIM: 604754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF271P PubMed Links

Gene Details

Gene
ZNF271P
Gene Name
zinc finger protein 271, pseudogene
There are no transcripts associated with this gene.

Gene
ZSCAN30
Gene Name
zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.

View Full Product Details