Product Details
- SNP ID
-
rs112279802
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:77841286 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCATGTCAGAGGTAGCAGCTTC[A/G]CATAAAAGCACCATTCCTAAGGTAA
- Phenotype
-
MIM: 300715
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGT1
PubMed Links
Gene Details
- Gene
- MAGT1
- Gene Name
- magnesium transporter 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032121.5 |
1019 |
Silent Mutation |
TGC,TGT |
C319C |
NP_115497.4 |
View Full Product Details