Product Details

SNP ID
rs117888525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85318514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACTCTTGCTCAAGCAAGTCA[C/T]GCCAAATCCAACCTTCCTAACAGGG
Phenotype
MIM: 604652 MIM: 603062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCF7L1 PubMed Links

Gene Details

Gene
TCF7L1
Gene Name
transcription factor 7 like 1
There are no transcripts associated with this gene.

Gene
TGOLN2
Gene Name
trans-golgi network protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206840.1 5696 UTR 3 NP_001193769.1
NM_001206841.1 5696 UTR 3 NP_001193770.1
NM_001206844.1 5696 UTR 3 NP_001193773.1
NM_006464.3 5696 UTR 3 NP_006455.2

View Full Product Details