Product Details

SNP ID
rs116629632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85321999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTTCTGTTTTATGCTCGGTAC[A/G]ACTAAGTGGAAGTTGATTCACTTTG
Phenotype
MIM: 603062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGOLN2 PubMed Links

Gene Details

Gene
TGOLN2
Gene Name
trans-golgi network protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206840.1 2211 UTR 3 NP_001193769.1
NM_001206841.1 2211 UTR 3 NP_001193770.1
NM_001206844.1 2211 UTR 3 NP_001193773.1
NM_006464.3 2211 UTR 3 NP_006455.2

View Full Product Details