Product Details
- SNP ID
-
rs115853953
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:174058180 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTGAGAGTGAGGTGGCTGGGGC[A/G]GGAGGGTCAAGGGTGAATGTTCAGT
- Phenotype
-
MIM: 616752
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HMP19
PubMed Links
Gene Details
- Gene
- HMP19
- Gene Name
- HMP19 protein
There are no transcripts associated with this gene.
View Full Product Details