Product Details

SNP ID
rs114880669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39039066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTAAAAATAAGCCCTGTTACCC[A/G]GCCCCCCCGCCCTTTTTTTGTTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NHLRC3 PubMed Links

Gene Details

Gene
NHLRC3
Gene Name
NHL repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012754.3 Intron NP_001012772.1
NM_001017370.2 Intron NP_001017370.1
Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 Intron NP_079414.3
XM_005266545.3 Intron XP_005266602.1
XM_011535239.2 Intron XP_011533541.1

View Full Product Details